Anti-WAS Mouse Monoclonal Antibody [clone: 336918]

Supplier: R & D Systems
MAB30701
220020-266EA 577.02 USD
220020-266
Anti-WAS Mouse Monoclonal Antibody [clone: 336918]
Antibodies
Type: Primary
Antigen: WAS
Clonality: Monoclonal
Clone: 336918
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR