Anti-WAS Mouse Monoclonal Antibody [clone: 336925]

Supplier: R & D Systems
MAB3070
220020-265EA 577.02 USD
220020-265
Anti-WAS Mouse Monoclonal Antibody [clone: 336925]
Antibodies
Type: Primary
Antigen: WAS
Clonality: Monoclonal
Clone: 336925
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR