Anti-IL1RAPL2 Rat Monoclonal Antibody [clone: 320017]

Supplier: R & D Systems
MAB3068
220020-264EA 577.02 USD
220020-264
Anti-IL1RAPL2 Rat Monoclonal Antibody [clone: 320017]
Antibodies
Type: Primary
Antigen: IL1RAPL2
Clonality: Monoclonal
Clone: 320017
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR