Anti-SORCS3 Rat Monoclonal Antibody [clone: 339615]

Supplier: R & D Systems
MAB30671
220020-263EA 577.02 USD
220020-263
Anti-SORCS3 Rat Monoclonal Antibody [clone: 339615]
Antibodies
Type: Primary
Antigen: SORCS3
Clonality: Monoclonal
Clone: 339615
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR