Anti-SORCS3 Rat Monoclonal Antibody [clone: 339624]

Supplier: R & D Systems
MAB3067
220020-262EA 594.37 USD
220020-262
Anti-SORCS3 Rat Monoclonal Antibody [clone: 339624]
Antibodies
Type: Primary
Antigen: SORCS3
Clonality: Monoclonal
Clone: 339624
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR