Anti-LILRB5 Mouse Monoclonal Antibody [clone: 395239]

Supplier: R & D Systems
MAB3065
220020-261EA 594.37 USD
220020-261
Anti-LILRB5 Mouse Monoclonal Antibody [clone: 395239]
Antibodies
Type: Primary
Antigen: LILRB5
Clonality: Monoclonal
Clone: 395239
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR