Anti-ITGB7 Rat Monoclonal Antibody [clone: 332428]

Supplier: R & D Systems
MAB30602
220020-259EA 594.37 USD
220020-259
Anti-ITGB7 Rat Monoclonal Antibody [clone: 332428]
Antibodies
Type: Primary
Antigen: ITGB7
Clonality: Monoclonal
Clone: 332428
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR