Anti-S100 A8 Rat Monoclonal Antibody [clone: 335806]

Supplier: R & D Systems
MAB3059
220020-255EA 639.2 USD
220020-255
Anti-S100 A8 Rat Monoclonal Antibody [clone: 335806]
Antibodies
Type: Primary
Antigen: S100 A8
Clonality: Monoclonal
Clone: 335806
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR