Anti-INSRR Mouse Monoclonal Antibody [clone: 326903]

Supplier: R & D Systems
MAB30551
220020-254EA 566.89 USD
220020-254
Anti-INSRR Mouse Monoclonal Antibody [clone: 326903]
Antibodies
Type: Primary
Antigen: INSRR
Clonality: Monoclonal
Clone: 326903
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR