Anti-INSRR Mouse Monoclonal Antibody [clone: 326929]

Supplier: R & D Systems
MAB3055
220020-253EA 566.89 USD
220020-253
Anti-INSRR Mouse Monoclonal Antibody [clone: 326929]
Antibodies
Type: Primary
Antigen: INSRR
Clonality: Monoclonal
Clone: 326929
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR