Anti-VCAN Rat Monoclonal Antibody [clone: 255915]

Supplier: R & D Systems
MAB3054
220020-252EA 763.57 USD
220020-252
Anti-VCAN Rat Monoclonal Antibody [clone: 255915]
Antibodies
Type: Primary
Antigen: VCAN
Clonality: Monoclonal
Clone: 255915
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR