Anti-VG5Q Rat Monoclonal Antibody [clone: 368011]

Supplier: R & D Systems
MAB30481
220020-249EA 561.11 USD
220020-249
Anti-VG5Q Rat Monoclonal Antibody [clone: 368011]
Antibodies
Type: Primary
Antigen: VG5Q
Clonality: Monoclonal
Clone: 368011
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR