Anti-IL32 Rat Monoclonal Antibody [clone: 373802]

Supplier: R & D Systems
MAB30401
102885-532EA 577.02 USD
102885-532
Anti-IL32 Rat Monoclonal Antibody [clone: 373802]
Antibodies
Type: Primary
Antigen: IL32
Clonality: Monoclonal
Clone: 373802
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR