Anti-EPHA2 Mouse Monoclonal Antibody [clone: 371805]

Supplier: R & D Systems
MAB3035
220020-243EA 655.11 USD
220020-243
Anti-EPHA2 Mouse Monoclonal Antibody [clone: 371805]
Antibodies
Type: Primary
Antigen: EPHA2
Clonality: Monoclonal
Clone: 371805
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR