Anti-EPHA1 Rat Monoclonal Antibody [clone: 404517]

Supplier: R & D Systems
MAB30341
220020-242EA 561.11 USD
220020-242
Anti-EPHA1 Rat Monoclonal Antibody [clone: 404517]
Antibodies
Type: Primary
Antigen: EPHA1
Clonality: Monoclonal
Clone: 404517
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR