Anti-EPHA1 Rat Monoclonal Antibody [clone: 404515]

Supplier: R & D Systems
MAB3034
220020-241EA 566.89 USD
220020-241
Anti-EPHA1 Rat Monoclonal Antibody [clone: 404515]
Antibodies
Type: Primary
Antigen: EPHA1
Clonality: Monoclonal
Clone: 404515
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR