Anti-CNTFR Mouse Monoclonal Antibody [clone: 121723]

Supplier: R & D Systems
MAB303
220020-240EA 772.25 USD
220020-240
Anti-CNTFR Mouse Monoclonal Antibody [clone: 121723]
Antibodies
Type: Primary
Antigen: CNTFR
Clonality: Monoclonal
Clone: 121723
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR