Anti-NEFM Mouse Monoclonal Antibody [clone: 327921]

Supplier: R & D Systems
MAB3029
220020-239EA 561.11 USD
220020-239
Anti-NEFM Mouse Monoclonal Antibody [clone: 327921]
Antibodies
Type: Primary
Antigen Symbol: NEFM
Antigen Name: neurofilament, medium polypeptide
Antigen Synonyms: NEF3, NF-M, NFM
Clonality: Monoclonal
Clone: 327921
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR