Anti-IL31 Rat Monoclonal Antibody [clone: 323407]

Supplier: R & D Systems
MAB3028
220020-238EA 633.42 USD
220020-238
Anti-IL31 Rat Monoclonal Antibody [clone: 323407]
Antibodies
Type: Primary
Antigen: IL31
Clonality: Monoclonal
Clone: 323407
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR