Anti-KEAP1 Mouse Monoclonal Antibody [clone: 333116]

Supplier: R & D Systems
MAB3024
220020-237EA 633.42 USD
220020-237
Anti-KEAP1 Mouse Monoclonal Antibody [clone: 333116]
Antibodies
Simple Western

Type: Primary
Antigen: KEAP1
Clonality: Monoclonal
Clone: 333116
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR