Anti-SAA1 Mouse Monoclonal Antibody [clone: 902738]

Supplier: R & D Systems
MAB30191
102885-530EA 594.37 USD
102885-530
Anti-SAA1 Mouse Monoclonal Antibody [clone: 902738]
Antibodies
Type: Primary
Antigen: SAA1
Clonality: Monoclonal
Clone: 902738
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR