Anti-SAA1 Rat Monoclonal Antibody [clone: 370820]

Supplier: R & D Systems
MAB3019
220020-236EA 616.06 USD
220020-236
Anti-SAA1 Rat Monoclonal Antibody [clone: 370820]
Antibodies
Type: Primary
Antigen: SAA1
Clonality: Monoclonal
Clone: 370820
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR