Anti-PGLYRP4 Mouse Monoclonal Antibody [clone: 474034]

Supplier: R & D Systems
MAB3018
220020-235EA 577.02 USD
220020-235
Anti-PGLYRP4 Mouse Monoclonal Antibody [clone: 474034]
Antibodies
Type: Primary
Antigen: PGLYRP4
Clonality: Monoclonal
Clone: 474034
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR