Anti-NDUFC2 Mouse Monoclonal Antibody [clone: 391934]

Supplier: R & D Systems
MAB3017
220020-234EA 584.25 USD
220020-234
Anti-NDUFC2 Mouse Monoclonal Antibody [clone: 391934]
Antibodies
Type: Primary
Antigen Symbol: NDUFC2
Antigen Name: NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa
Antigen Synonyms: B14.5b, CI-B14.5b, HLC-1, NADHDH2
Clonality: Monoclonal
Clone: 391934
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR