Anti-NDP Mouse Monoclonal Antibody [clone: 343711]

Supplier: R & D Systems
MAB3014
220020-233EA 577.02 USD
220020-233
Anti-NDP Mouse Monoclonal Antibody [clone: 343711]
Antibodies
Type: Primary
Antigen Symbol: NDP
Antigen Name: Norrie disease (pseudoglioma)
Antigen Synonyms: EVR2, FEVR, ND
Clonality: Monoclonal
Clone: 343711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR