Anti-SLITRK1 Mouse Monoclonal Antibody [clone: 327808]

Supplier: R & D Systems
MAB3009
220020-229EA 577.02 USD
220020-229
Anti-SLITRK1 Mouse Monoclonal Antibody [clone: 327808]
Antibodies
Type: Primary
Antigen: SLITRK1
Clonality: Monoclonal
Clone: 327808
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR