Anti-TLR3 Rat Monoclonal Antibody [clone: 313129]

Supplier: R & D Systems
MAB3005
220020-227EA 584.25 USD
220020-227
Anti-TLR3 Rat Monoclonal Antibody [clone: 313129]
Antibodies
Type: Primary
Antigen: TLR3
Clonality: Monoclonal
Clone: 313129
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR