Anti-PPP1CA Mouse Monoclonal Antibody [clone: 319319]

Supplier: R & D Systems
MAB3000
220020-225EA 577.02 USD
220020-225
Anti-PPP1CA Mouse Monoclonal Antibody [clone: 319319]
Antibodies
Type: Primary
Antigen: PPP1CA
Clonality: Monoclonal
Clone: 319319
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR