Anti-PIK3CG Rat Monoclonal Antibody [clone: 304103]

Supplier: R & D Systems
MAB2999
220020-224EA 577.02 USD
220020-224
Anti-PIK3CG Rat Monoclonal Antibody [clone: 304103]
Antibodies
Type: Primary
Antigen: PIK3CG
Clonality: Monoclonal
Clone: 304103
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR