Anti-PIK3R1 Mouse Monoclonal Antibody [clone: 572106]

Supplier: R & D Systems
MAB2998
220020-223EA 594.37 USD
220020-223
Anti-PIK3R1 Mouse Monoclonal Antibody [clone: 572106]
Antibodies
Simple Western

Type: Primary
Antigen: PIK3R1
Clonality: Monoclonal
Clone: 572106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR