Anti-GSK3A Mouse Monoclonal Antibody [clone: 288901]

Supplier: R & D Systems
MAB2986
220020-220EA 566.89 USD
220020-220
Anti-GSK3A Mouse Monoclonal Antibody [clone: 288901]
Antibodies
Type: Primary
Antigen Symbol: GSK3A
Antigen Name: glycogen synthase kinase 3 alpha
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 288901
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR