Anti-SERPINE2 Mouse Monoclonal Antibody [clone: 449888]

Supplier: R & D Systems
MAB2980
220020-219EA 577.02 USD
220020-219
Anti-SERPINE2 Mouse Monoclonal Antibody [clone: 449888]
Antibodies
Type: Primary
Antigen: SERPINE2
Clonality: Monoclonal
Clone: 449888
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR