Anti-TFPI Rat Monoclonal Antibody [clone: 372311]

Supplier: R & D Systems
MAB2975
220020-218EA 577.02 USD
220020-218
Anti-TFPI Rat Monoclonal Antibody [clone: 372311]
Antibodies
Type: Primary
Antigen: TFPI
Clonality: Monoclonal
Clone: 372311
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR