Anti-TFPI Mouse Monoclonal Antibody [clone: 374718]

Supplier: R & D Systems
MAB29741
220020-217EA 772.25 USD
220020-217
Anti-TFPI Mouse Monoclonal Antibody [clone: 374718]
Antibodies
Type: Primary
Antigen: TFPI
Clonality: Monoclonal
Clone: 374718
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR