Anti-TFPI Mouse Monoclonal Antibody [clone: 374720]

Supplier: R & D Systems
MAB2974
220020-216EA 577.02 USD
220020-216
Anti-TFPI Mouse Monoclonal Antibody [clone: 374720]
Antibodies
Type: Primary
Antigen: TFPI
Clonality: Monoclonal
Clone: 374720
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR