Anti-XPNPEP1 Mouse Monoclonal Antibody [clone: 409111]

Supplier: R & D Systems
MAB2970
220020-214EA 577.02 USD
220020-214
Anti-XPNPEP1 Mouse Monoclonal Antibody [clone: 409111]
Antibodies
Type: Primary
Antigen: XPNPEP1
Clonality: Monoclonal
Clone: 409111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR