Anti-VEGF Mouse Monoclonal Antibody [clone: 146213]

Supplier: R & D Systems
MAB297
220020-213EA 763.57 USD
220020-213
Anti-VEGF Mouse Monoclonal Antibody [clone: 146213]
Antibodies
Type: Primary
Antigen: VEGF
Clonality: Monoclonal
Clone: 146213
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR