Anti-PRSS8 Rat Monoclonal Antibody [clone: 430814]

Supplier: R & D Systems
MAB2968
220020-212EA 561.11 USD
220020-212
Anti-PRSS8 Rat Monoclonal Antibody [clone: 430814]
Antibodies
Type: Primary
Antigen: PRSS8
Clonality: Monoclonal
Clone: 430814
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR