Anti-MMP7 Rat Monoclonal Antibody [clone: 377313]

Supplier: R & D Systems
MAB29671
220020-211EA 577.02 USD
220020-211
Anti-MMP7 Rat Monoclonal Antibody [clone: 377313]
Antibodies
Type: Primary
Antigen: MMP7
Clonality: Monoclonal
Clone: 377313
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR