Anti-SDC1 Rat Monoclonal Antibody [clone: 320611]

Supplier: R & D Systems
MAB29661
220020-210EA 577.02 USD
220020-210
Anti-SDC1 Rat Monoclonal Antibody [clone: 320611]
Antibodies
Type: Primary
Antigen: SDC1
Clonality: Monoclonal
Clone: 320611
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR