Anti-SDC1 Rat Monoclonal Antibody [clone: 300506]

Supplier: R & D Systems
MAB2966
220020-209EA 594.37 USD
220020-209
Anti-SDC1 Rat Monoclonal Antibody [clone: 300506]
Antibodies
Type: Primary
Antigen: SDC1
Clonality: Monoclonal
Clone: 300506
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR