Anti-SDC2 Rat Monoclonal Antibody [clone: 305515]

Supplier: R & D Systems
MAB2965
220020-207EA 594.37 USD
220020-207
Anti-SDC2 Rat Monoclonal Antibody [clone: 305515]
Antibodies
Type: Primary
Antigen: SDC2
Clonality: Monoclonal
Clone: 305515
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR