Anti-FABP1 Mouse Monoclonal Antibody [clone: 328605]

Supplier: R & D Systems
MAB29641
220020-206EA 594.37 USD
220020-206
Anti-FABP1 Mouse Monoclonal Antibody [clone: 328605]
Antibodies
Simple Western

Type: Primary
Antigen Symbol: FABP1
Antigen Name: fatty acid binding protein 1, liver
Antigen Synonyms: FABPL, L-FABP
Clonality: Monoclonal
Clone: 328605
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR