Anti-LRP6 Rat Monoclonal Antibody [clone: 290913]

Supplier: R & D Systems
MAB2960
220020-204EA 594.37 USD
220020-204
Anti-LRP6 Rat Monoclonal Antibody [clone: 290913]
Antibodies
Type: Primary
Antigen: LRP6
Clonality: Monoclonal
Clone: 290913
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR