Anti-SUMO3 Rat Monoclonal Antibody [clone: 401513]

Supplier: R & D Systems
MAB2959
220020-203EA 577.02 USD
220020-203
Anti-SUMO3 Rat Monoclonal Antibody [clone: 401513]
Antibodies
Type: Primary
Antigen: SUMO3
Clonality: Monoclonal
Clone: 401513
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR