Anti-STC1 Mouse Monoclonal Antibody [clone: 380715]

Supplier: R & D Systems
MAB2958
220020-202EA 594.37 USD
220020-202
Anti-STC1 Mouse Monoclonal Antibody [clone: 380715]
Antibodies
Type: Primary
Antigen: STC1
Clonality: Monoclonal
Clone: 380715
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR