Anti-S100 P Mouse Monoclonal Antibody [clone: 357517]

Supplier: R & D Systems
MAB2957
220020-201EA 594.37 USD
220020-201
Anti-S100 P Mouse Monoclonal Antibody [clone: 357517]
Antibodies
Type: Primary
Antigen: S100 P
Clonality: Monoclonal
Clone: 357517
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR