Anti-POSTN Rat Monoclonal Antibody [clone: 345616]

Supplier: R & D Systems
MAB2955
220020-200EA 763.57 USD
220020-200
Anti-POSTN Rat Monoclonal Antibody [clone: 345616]
Antibodies
Type: Primary
Antigen: POSTN
Clonality: Monoclonal
Clone: 345616
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR