Anti-OTUD4 Rat Monoclonal Antibody [clone: 368203]

Supplier: R & D Systems
MAB2954
220020-199EA 577.02 USD
220020-199
Anti-OTUD4 Rat Monoclonal Antibody [clone: 368203]
Antibodies
Type: Primary
Antigen: OTUD4
Clonality: Monoclonal
Clone: 368203
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR