Anti-OSM Mouse Monoclonal Antibody [clone: 17022]

Supplier: R & D Systems
MAB2951
102885-528EA 763.57 USD
102885-528
Anti-OSM Mouse Monoclonal Antibody [clone: 17022]
Antibodies
Type: Primary
Antigen: OSM
Clonality: Monoclonal
Clone: 17022
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR